Learn More
Medchemexpress LLC M-PEG12-azide | 2170098-29-8 | >98.6% | C25H51N3O12 | 5 G
SDP
Catalog No. 5000353384
Encompass_Preferred
m-PEG12-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates various conjugation reactions.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-53384 Supplier Medchemexpress LLC Supplier No. HY1408305G
m-PEG12-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates various conjugation reactions.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups