Learn More
Medchemexpress LLC M-PEG12-DBCO | 96.9% | C44H66N2O14 | 50 MG
SDP
Catalog No. 5000355044
Encompass_Preferred
m-PEG12-DBCO is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins
Catalog No. 50-003-55044 Supplier Medchemexpress LLC Supplier No. HY14031750MG
m-PEG12-DBCO is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins