Learn More
Medchemexpress LLC M-PEG13-acid | 2170098-33-4 | 99.9% | 632.74 g/mol | C28H56O15 | 5 G
SDP
Catalog No. 5000281935
Encompass_Preferred
m-PEG13-acid is a methoxy polyethylene glycol (mPEG) linker bearing a terminal carboxylic acid. It functions as a hydrophilic spacer for bioconjugation and biochemical assays; the terminal carboxyl can be activated (for example with EDC or HATU) and coupled to primary amines to form stable amide bonds, while the PEG spacer enhances aqueous solubility.
Catalog No. 50-002-81935 Supplier Medchemexpress LLC Supplier No. HYW1907525G
m-PEG13-acid is a methoxy polyethylene glycol (mPEG) linker bearing a terminal carboxylic acid. It functions as a hydrophilic spacer for bioconjugation and biochemical assays; the terminal carboxyl can be activated (for example with EDC or HATU) and coupled to primary amines to form stable amide bonds, while the PEG spacer enhances aqueous solubility.