Learn More
Medchemexpress LLC M-PEG13-acid | 2170098-33-4 | MFCD28385486 | >95.0% | 632.74 g·mol⁻¹ | C28H56O15 | 100 MG
SDP
Catalog No. 5000281931
Encompass_Preferred
m-PEG13-acid is a polyethylene glycol (PEG) linker bearing a terminal carboxylic acid used in bioconjugation and PEGylation chemistry. It provides a hydrophilic spacer that facilitates formation of amide bonds to primary amines after activation, improving solubility and flexibility of conjugates. Typical identifiers: molecular weight ~632.74 g·mol⁻¹, formula C28H56O15, CAS 2170098-33-4, purity >95.0%.
  • Acts as a hydrophilic spacer for bioconjugation.
  • Contains a terminal carboxylic acid for amide bond formation with primary amines.
  • Improves solubility and flexibility of conjugated molecules.
  • Compatible with common coupling agents such as EDC and HATU.
  • Supplied at >95.0% purity for research applications.
Catalog No. 50-002-81931 Supplier Medchemexpress LLC Supplier No. HYW190752100MG
m-PEG13-acid is a polyethylene glycol (PEG) linker bearing a terminal carboxylic acid used in bioconjugation and PEGylation chemistry. It provides a hydrophilic spacer that facilitates formation of amide bonds to primary amines after activation, improving solubility and flexibility of conjugates. Typical identifiers: molecular weight ~632.74 g·mol⁻¹, formula C28H56O15, CAS 2170098-33-4, purity >95.0%.
  • Acts as a hydrophilic spacer for bioconjugation.
  • Contains a terminal carboxylic acid for amide bond formation with primary amines.
  • Improves solubility and flexibility of conjugated molecules.
  • Compatible with common coupling agents such as EDC and HATU.
  • Supplied at >95.0% purity for research applications.