Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG13-NHS ester | 3063847-98-0 | 95.0% | C32H59NO17 | 25 MG
SDP

Catalog No. 5000352788
Encompass_Preferred

M-PEG13-NHS ester is a PEG-based PROTAC linker, useful in the synthesis of PROTACs. This compound appears as a colorless to off-white oil.

  • PEG-based PROTAC linker
  • Suitable for synthesis of PROTACs
  • Appearance: Colorless to off-white oil
  • Purity: 95.0%
  • Molecular weight: 729.81
  • CAS number: 3063847-98-0

Catalog No. 50-003-52788 Supplier Medchemexpress LLC Supplier No. 25MGHY141107
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M-PEG13-NHS ester is a PEG-based PROTAC linker, useful in the synthesis of PROTACs. This compound appears as a colorless to off-white oil.

  • PEG-based PROTAC linker
  • Suitable for synthesis of PROTACs
  • Appearance: Colorless to off-white oil
  • Purity: 95.0%
  • Molecular weight: 729.81
  • CAS number: 3063847-98-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.