Learn More
Medchemexpress LLC M-PEG14-NHS ester | 00-00-0 | 95.0% | 773.86 g/mol | C34H63NO18 | 5 MG
SDP
Catalog No. 5000222986
Encompass_Preferred
m-PEG14-NHS ester is a methoxy-terminated polyethylene glycol linker bearing an N-hydroxysuccinimide (NHS) ester functional group. It is designed for amine-reactive conjugation in the synthesis of proteolysis targeting chimeras (PROTACs) and related bioconjugation workflows, supplied as a white to off-white solid for research use only.
  • Provides an m-PEG14 spacer for flexible linker length
  • NHS ester enables efficient coupling to primary amines
  • High purity suitable for synthetic applications
  • Offered in small quantities for laboratory-scale synthesis
  • Stable when stored sealed at low temperature and protected from moisture
Catalog No. 50-002-22986 Supplier Medchemexpress LLC Supplier No. HY1387185MG
m-PEG14-NHS ester is a methoxy-terminated polyethylene glycol linker bearing an N-hydroxysuccinimide (NHS) ester functional group. It is designed for amine-reactive conjugation in the synthesis of proteolysis targeting chimeras (PROTACs) and related bioconjugation workflows, supplied as a white to off-white solid for research use only.
  • Provides an m-PEG14 spacer for flexible linker length
  • NHS ester enables efficient coupling to primary amines
  • High purity suitable for synthetic applications
  • Offered in small quantities for laboratory-scale synthesis
  • Stable when stored sealed at low temperature and protected from moisture