Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG14-NHS ester | 00-00-0 | 95.0% | C34H63NO18 | 10MG
SDP

Catalog No. 5000260681
Encompass_Preferred

m-PEG14-NHS ester is a methoxy-terminated polyethylene glycol (PEG) reagent bearing an N-hydroxysuccinimide (NHS) active ester. Supplied as a solid for laboratory use, it is intended for use as a PROTAC linker or for PEGylation to attach PEG chains to amine-containing molecules via stable amide bonds.

  • Methoxy-terminated PEG with an NHS active ester.
  • Reacts with primary amines to form stable amide bonds.
  • Molecular weight ~773.86 Da.
  • Chemical formula C34H63NO18.
  • Purity ≥95.0%.
  • White to off-white solid appearance.
  • Store sealed, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in small research pack sizes.

Catalog No. 50-002-60681 Supplier Medchemexpress LLC Supplier No. HY13871810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG14-NHS ester is a methoxy-terminated polyethylene glycol (PEG) reagent bearing an N-hydroxysuccinimide (NHS) active ester. Supplied as a solid for laboratory use, it is intended for use as a PROTAC linker or for PEGylation to attach PEG chains to amine-containing molecules via stable amide bonds.

  • Methoxy-terminated PEG with an NHS active ester.
  • Reacts with primary amines to form stable amide bonds.
  • Molecular weight ~773.86 Da.
  • Chemical formula C34H63NO18.
  • Purity ≥95.0%.
  • White to off-white solid appearance.
  • Store sealed, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Available in small research pack sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.