Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG14-NHS ester | 95.0% | 773.86 | 25 MG
SDP

Catalog No. 5000348585
Encompass_Preferred

m-PEG14-NHS ester is a PEG-based PROTAC linker. This chemical is intended for laboratory use in the synthesis of PROTACs.

  • Supplied as a white to off-white solid
  • Stable under recommended storage conditions
  • Recommended storage at -20°C, sealed storage, away from moisture and light
  • Shipping at room temperature if less than 2 weeks
  • Has a purity of 95.0%
  • Molecular weight is 773.86

Catalog No. 50-003-48585 Supplier Medchemexpress LLC Supplier No. 25MGHY138718
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG14-NHS ester is a PEG-based PROTAC linker. This chemical is intended for laboratory use in the synthesis of PROTACs.

  • Supplied as a white to off-white solid
  • Stable under recommended storage conditions
  • Recommended storage at -20°C, sealed storage, away from moisture and light
  • Shipping at room temperature if less than 2 weeks
  • Has a purity of 95.0%
  • Molecular weight is 773.86

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.