Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG15-amine | 2745717-94-4 | 99.9% | 691.85 | 25 MG
SDP

Catalog No. 5000348647
Encompass_Preferred

m-PEG15-amine (CAS No.: 2745717-94-4) is a high-purity PEG-based PROTAC linker with a molecular formula of C31H65NO15 and a molecular weight of 691.85.

  • High purity of 99.9%
  • Appears as a colorless to light yellow oil
  • Store at -20°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-48647 Supplier Medchemexpress LLC Supplier No. 25MGHY138424
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG15-amine (CAS No.: 2745717-94-4) is a high-purity PEG-based PROTAC linker with a molecular formula of C31H65NO15 and a molecular weight of 691.85.

  • High purity of 99.9%
  • Appears as a colorless to light yellow oil
  • Store at -20°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.