Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG15-NHS ester | 00-00-0 | 98.0% | C39H73NO20 | 10MG
SDP

Catalog No. 5000258892
Encompass_Preferred

m-PEG15-NHS ester is a methoxy-terminated polyethylene glycol (PEG) N-hydroxysuccinimide ester used as a hydrophilic linker in PROTAC synthesis and bioconjugation. It supplies an amine-reactive NHS group for covalent coupling to primary amines and a PEG15 spacer to improve solubility, flexibility, and distance between conjugated moieties.

  • Provides an NHS ester for efficient amine coupling.
  • Contains a methoxy-terminated PEG15 spacer to increase solubility.
  • High purity (98.0%) suitable for research applications.
  • Storage recommended at -20°C, sealed and protected from moisture and light.
  • Available in multiple package sizes to support different scales of synthesis.

Catalog No. 50-002-58892 Supplier Medchemexpress LLC Supplier No. HY13082910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG15-NHS ester is a methoxy-terminated polyethylene glycol (PEG) N-hydroxysuccinimide ester used as a hydrophilic linker in PROTAC synthesis and bioconjugation. It supplies an amine-reactive NHS group for covalent coupling to primary amines and a PEG15 spacer to improve solubility, flexibility, and distance between conjugated moieties.

  • Provides an NHS ester for efficient amine coupling.
  • Contains a methoxy-terminated PEG15 spacer to increase solubility.
  • High purity (98.0%) suitable for research applications.
  • Storage recommended at -20°C, sealed and protected from moisture and light.
  • Available in multiple package sizes to support different scales of synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.