Learn More
Medchemexpress LLC m-PEG15-NHS ester | 98.0% | 25 MG
SDP
Catalog No. 5000342290
Encompass_Preferred
m-PEG15-NHS ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs degrade target proteins via the ubiquitin-proteasome system
  • PROTACs utilize two ligands connected by a linker for E3 ubiquitin ligase and target protein
Catalog No. 50-003-42290 Supplier Medchemexpress LLC Supplier No. 25MGHY130829
m-PEG15-NHS ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs degrade target proteins via the ubiquitin-proteasome system
  • PROTACs utilize two ligands connected by a linker for E3 ubiquitin ligase and target protein