Learn More
Medchemexpress LLC m-PEG15-NHS ester | 98.0% | 50 MG
SDP
Catalog No. 5000338990
Encompass_Preferred
m-PEG15-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
Catalog No. 50-003-38990 Supplier Medchemexpress LLC Supplier No. HY13082950MG
m-PEG15-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other a target protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs