Learn More
Medchemexpress LLC M-PEG15-NHS ester | 98.0% | 875.99 g/mol | C39H73NO20 | 5 MG
SDP
Catalog No. 5000262912
Encompass_Preferred
m-PEG15-NHS ester is a polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester for amine-specific conjugation. It provides a PEG15 spacer to increase solubility and flexibility during bioconjugation and linker assembly, and is supplied with defined purity and molecular weight for research applications.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • PEG15 spacer to enhance solubility and reduce steric hindrance.
  • High purity suitable for research-grade bioconjugation.
  • Stable when stored sealed at -20°C to preserve activity.
  • Designed for linker synthesis and PROTAC construction workflows.
Catalog No. 50-002-62912 Supplier Medchemexpress LLC Supplier No. HY1308295MG
m-PEG15-NHS ester is a polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) ester for amine-specific conjugation. It provides a PEG15 spacer to increase solubility and flexibility during bioconjugation and linker assembly, and is supplied with defined purity and molecular weight for research applications.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • PEG15 spacer to enhance solubility and reduce steric hindrance.
  • High purity suitable for research-grade bioconjugation.
  • Stable when stored sealed at -20°C to preserve activity.
  • Designed for linker synthesis and PROTAC construction workflows.