Learn More
Medchemexpress LLC M-PEG16-COOH | 2933174-52-6 | 98.0% | 764.89 | 100 MG
SDP
Catalog No. 5000349442
Encompass_Preferred
M-PEG16-COOH is a PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs, which are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Can be used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49442 Supplier Medchemexpress LLC Supplier No. HY138314100MG
Add to Cart
Add to Cart
M-PEG16-COOH is a PEG-based PROTAC linker. It is designed for use in the synthesis of PROTACs, which are compounds that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Can be used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins