Learn More
Medchemexpress LLC m-PEG16-NHS ester | 2563873-83-4 | 100mg
SDP
Catalog No. 5000270233
Encompass_Preferred
m-PEG16-NHS ester is a PEG derivative composed of an NHS ester that can be conjugated to amino acids or other molecules containing amino groups[1]
Catalog No. 50-002-70233 Supplier Medchemexpress LLC Supplier No. HYW879004100MG
m-PEG16-NHS ester is a PEG derivative composed of an NHS ester that can be conjugated to amino acids or other molecules containing amino groups[1]