Learn More
Medchemexpress LLC M-PEG16-NHS ester | 2563873-83-4 | MFCD13184971 | ≥97.0% | 861.97 | C38H71NO20 | 250mg
SDP
Supplier:  Medchemexpress LLC HYW879004250MG
Encompass_Preferred
m-PEG16-NHS ester is a PEG derivative composed of an NHS ester that can be conjugated to amino acids or other molecules containing amino groups[1]
Catalog No. 50-002-68369