Learn More
Medchemexpress LLC M-PEG17-acid | 2346581-96-0 | 98.5% | C36H72O19 | 100 MG
SDP
Catalog No. 5000353068
Encompass_Preferred
m-PEG17-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules employ the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system.
Catalog No. 50-003-53068 Supplier Medchemexpress LLC Supplier No. HY140502100MG
m-PEG17-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules employ the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Facilitates selective degradation of target proteins via the ubiquitin-proteasome system.