Learn More
Medchemexpress LLC M-PEG17-acid | 2346581-96-0 | 98.5% | C36H72O19 | 50 MG
SDP
Catalog No. 5000353226
Encompass_Preferred
m-PEG17-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-53226 Supplier Medchemexpress LLC Supplier No. HY14050250MG
m-PEG17-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.