Learn More
Medchemexpress LLC M-PEG2-acid | 149577-05-9 | 25 G
Supplier: Medchemexpress LLC 25GHY130231
m-PEG2-acid is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteeasome system to selectively degrade target proteins.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Consists of two distinct ligands connected by a linker
- Targets an E3 ubiquitin ligase
- Targets a specific protein
- Harnesses the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins
- For research use only