Learn More
Medchemexpress LLC M-PEG2-acid | 149577-05-9 | 5 G
SDP
Catalog No. 5000338581
Encompass_Preferred
m-PEG2-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. These molecules typically consist of two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation of target proteins
  • Intended for research use only
Catalog No. 50-003-38581 Supplier Medchemexpress LLC Supplier No. HY1302315G
m-PEG2-acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. These molecules typically consist of two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other binds to the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system for selective degradation of target proteins
  • Intended for research use only