Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG2-acid | 149577-05-9 | >98% | 1 G
SDP

Catalog No. 5000338509
Encompass_Preferred

m-PEG2-acid is a polyethylene glycol (PEG)-based PROTAC linker. It is primarily used in the synthesis of Proteolysis-Targeting Chimeras (PROTACs), which are molecules designed to exploit the intracellular ubiquitin-proteasome system. PROTACs achieve selective degradation of target proteins by linking them to an E3 ubiquitin ligase.

  • PEG-based PROTAC linker
  • Facilitates the synthesis of PROTACs
  • Molecular weight of 148.16
  • Chemical formula C6H12O4
  • Appears as a colorless to light yellow liquid
  • Density of 1.098 ±0.06 g/cm³
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-38509 Supplier Medchemexpress LLC Supplier No. HY1302311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG2-acid is a polyethylene glycol (PEG)-based PROTAC linker. It is primarily used in the synthesis of Proteolysis-Targeting Chimeras (PROTACs), which are molecules designed to exploit the intracellular ubiquitin-proteasome system. PROTACs achieve selective degradation of target proteins by linking them to an E3 ubiquitin ligase.

  • PEG-based PROTAC linker
  • Facilitates the synthesis of PROTACs
  • Molecular weight of 148.16
  • Chemical formula C6H12O4
  • Appears as a colorless to light yellow liquid
  • Density of 1.098 ±0.06 g/cm³
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.