Learn More
Medchemexpress LLC m-PEG2-azide | 215181-61-6 | 250 MG
SDP
Catalog No. 5000343016
Encompass_Preferred
m-PEG2-azide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It serves as a click chemistry reagent, featuring an Azide group. This enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only
Catalog No. 50-003-43016 Supplier Medchemexpress LLC Supplier No. 250MGHY130578
m-PEG2-azide is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It serves as a click chemistry reagent, featuring an Azide group. This enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • For research use only