Learn More
Medchemexpress LLC M-PEG2-azide | 215181-61-6 | 5 G
SDP
Catalog No. 5000340618
Encompass_Preferred
m-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • Click chemistry reagent containing an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-40618 Supplier Medchemexpress LLC Supplier No. HY1305785G
m-PEG2-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs.
  • Click chemistry reagent containing an azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups