Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG2-CH2CH2COOH | 209542-49-4 | 192.21 g/mol | C8H16O5 | 1 G
SDP

Catalog No. 5000210206
Encompass_Preferred

m-PEG2-CH2CH2COOH is a PEG-based PROTAC linker used to connect target-binding ligands and E3 ligase ligands in PROTAC molecules. It is supplied as a research reagent for chemical synthesis and proteolysis-targeting chimera development.

  • PEG-based linker for PROTAC synthesis
  • CAS number 209542-49-4
  • Molecular weight 192.21 g/mol
  • Chemical formula C8H16O5
  • Available in multiple sizes, including 1 G
  • For research use only, not for human or clinical use

Catalog No. 50-002-10206 Supplier Medchemexpress LLC Supplier No. HYW0437251G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG2-CH2CH2COOH is a PEG-based PROTAC linker used to connect target-binding ligands and E3 ligase ligands in PROTAC molecules. It is supplied as a research reagent for chemical synthesis and proteolysis-targeting chimera development.

  • PEG-based linker for PROTAC synthesis
  • CAS number 209542-49-4
  • Molecular weight 192.21 g/mol
  • Chemical formula C8H16O5
  • Available in multiple sizes, including 1 G
  • For research use only, not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.