Learn More
Medchemexpress LLC M-PEG2-MS | 60696-83-5 | C6H14O5S | 250 MG
SDP
Catalog No. 5000352726
Encompass_Preferred
m-PEG2-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.
Catalog No. 50-003-52726 Supplier Medchemexpress LLC Supplier No. 250MGHY140362
m-PEG2-MS is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PROTACs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase.
  • The other ligand is for the target protein.