Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG2-Tos | 50586-80-6 | 274.33 g/mol | C12H18O5S | 10 G
SDP

Catalog No. 5000202336
Encompass_Preferred

m-PEG2-Tos is a PEG-derived, uncleavable linker used for bioconjugation in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs). It provides a short PEG spacer that improves solubility and stability between the targeting moiety and payload.

  • Uncleavable PEG-based linker suitable for ADC and PROTAC construction.
  • Short PEG spacer improves solubility and reduces aggregation.
  • Molecular weight 274.33 g/mol, formula C12H18O5S.
  • CAS number 50586-80-6.
  • Available in laboratory pack sizes, including 10 g.

Catalog No. 50-002-02336 Supplier Medchemexpress LLC Supplier No. HY4274510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG2-Tos is a PEG-derived, uncleavable linker used for bioconjugation in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs). It provides a short PEG spacer that improves solubility and stability between the targeting moiety and payload.

  • Uncleavable PEG-based linker suitable for ADC and PROTAC construction.
  • Short PEG spacer improves solubility and reduces aggregation.
  • Molecular weight 274.33 g/mol, formula C12H18O5S.
  • CAS number 50586-80-6.
  • Available in laboratory pack sizes, including 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.