Learn More
Medchemexpress LLC m-PEG20-alcohol 250mg
SDP
Catalog No. 5000204745
Encompass_Preferred
m-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04745 Supplier Medchemexpress LLC Supplier No. 250MGHYW096138
m-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]