Learn More
Medchemexpress LLC HY-133323 100mg Medchemexpress, m-PEG24-acid CAS:2248203-61-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133323100MG
Encompass_Preferred
Medchemexpress, HY-133323 100mg m-PEG24-acid CAS:2248203-61-2 m-PEG24-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1337