Learn More
Medchemexpress LLC M-PEG24-alcohol | 2376450-73-4 | 99.7% | C49H100O25 | 250 MG
SDP
Catalog No. 5000352727
Encompass_Preferred
m-PEG24-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins by utilizing two ligands connected by this linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
Catalog No. 50-003-52727 Supplier Medchemexpress LLC Supplier No. 250MGHY141225
m-PEG24-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins by utilizing two ligands connected by this linker: one for an E3 ubiquitin ligase and the other for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation