Learn More
Medchemexpress LLC M-PEG24-alcohol | 2376450-73-4 | MFCD27977524 | C49H100O25 | 10MG
SDP
Catalog No. 5000260901
Encompass_Preferred
m-PEG24-alcohol is a methoxy-terminated polyethylene glycol (PEG) linker used as a hydrophilic spacer in the synthesis of bifunctional degrader molecules such as PROTACs. The 24-unit PEG chain provides flexibility and solubility to tune linker length, target engagement, and pharmacokinetic properties; consult the certificate of analysis for purity and recommended storage conditions.
  • Acts as a hydrophilic, flexible spacer between ligands.
  • Helps modulate ternary complex formation in degrader design.
  • Improves solubility and linker conformational freedom.
  • Available in small research-scale quantities for synthetic work.
  • Certificate of analysis provides purity and storage information.
Catalog No. 50-002-60901 Supplier Medchemexpress LLC Supplier No. HY14122510MG
m-PEG24-alcohol is a methoxy-terminated polyethylene glycol (PEG) linker used as a hydrophilic spacer in the synthesis of bifunctional degrader molecules such as PROTACs. The 24-unit PEG chain provides flexibility and solubility to tune linker length, target engagement, and pharmacokinetic properties; consult the certificate of analysis for purity and recommended storage conditions.
  • Acts as a hydrophilic, flexible spacer between ligands.
  • Helps modulate ternary complex formation in degrader design.
  • Improves solubility and linker conformational freedom.
  • Available in small research-scale quantities for synthetic work.
  • Certificate of analysis provides purity and storage information.