Learn More
Medchemexpress LLC m-PEG24-NHS ester | 2395839-96-8 | 99.7% | 25 MG
SDP
Catalog No. 5000339498
Encompass_Preferred
m-PEG24-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins, offering a novel approach in therapeutic development. This product is intended for research use only and is supplied as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Stable under recommended storage conditions
Catalog No. 50-003-39498 Supplier Medchemexpress LLC Supplier No. 25MGHY130830
m-PEG24-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins, offering a novel approach in therapeutic development. This product is intended for research use only and is supplied as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Stable under recommended storage conditions