Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG24-NHS ester | 2395839-96-8 | 99.7% | 50 MG
SDP

Catalog No. 5000339001
Encompass_Preferred

m-PEG24-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. They harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Appearance: White to off-white solid
  • Purity (HPLC): 99.7%

Catalog No. 50-003-39001 Supplier Medchemexpress LLC Supplier No. HY13083050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG24-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. They harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Appearance: White to off-white solid
  • Purity (HPLC): 99.7%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.