Learn More
Medchemexpress LLC m-PEG24-NHS ester | 2395839-96-8 | 99.9% | 100 MG
SDP
Catalog No. 5000340211
Encompass_Preferred
m-PEG24-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 1214.39 and the chemical formula C54H103NO28, appearing as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • High purity of 99.9%
  • White to off-white solid appearance
Catalog No. 50-003-40211 Supplier Medchemexpress LLC Supplier No. HY130830100MG
m-PEG24-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 1214.39 and the chemical formula C54H103NO28, appearing as a white to off-white solid.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • High purity of 99.9%
  • White to off-white solid appearance