Learn More
Medchemexpress LLC M-PEG3-0-benzaldehyde | 153364-63-7 | >98% | C14H20O5 | 1 G
SDP
Catalog No. 5000353352
Encompass_Preferred
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
Catalog No. 50-003-53352 Supplier Medchemexpress LLC Supplier No. HY1406211G
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.