Learn More
Medchemexpress LLC M-peg3-0-benzaldehyde | 153364-63-7 | C14H20O5 | 250 MG
SDP
Catalog No. 5000357250
Encompass_Preferred
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and is not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Selectively degrade target proteins
  • For research use only
Catalog No. 50-003-57250 Supplier Medchemexpress LLC Supplier No. 250MGHY140621
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only and is not sold to patients.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Selectively degrade target proteins
  • For research use only