Learn More
Medchemexpress LLC M-PEG3-0-benzaldehyde | 153364-63-7 | C14H20O5 | 5 G
SDP
Catalog No. 5000356449
Encompass_Preferred
M-PEG3-0-benzaldehyde is a PEG-based PROTAC linker. PROTACs contain two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-56449 Supplier Medchemexpress LLC Supplier No. HY1406215G
M-PEG3-0-benzaldehyde is a PEG-based PROTAC linker. PROTACs contain two different ligands connected by a linker, with one ligand for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes the ubiquitin-proteasome system for selective protein degradation