Learn More
Medchemexpress LLC M-PEG3-0-benzaldehyde | 153364-63-7 | C14H20O5 | 500 MG
SDP
Catalog No. 5000355641
Encompass_Preferred
M-PEG3-0-benzaldehyde is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other targeting a specific protein. These molecules utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55641 Supplier Medchemexpress LLC Supplier No. HY140621500MG
M-PEG3-0-benzaldehyde is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other targeting a specific protein. These molecules utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins