Learn More
Medchemexpress LLC m-PEG3-aldehyde | 356066-46-1 | MFCD26910955 | 99.8% | 176.21 | C8H16O4 | 100MG
SDP
Catalog No. 5000235035
Encompass_Preferred
m-PEG3-aldehyde is a monomethyl triethylene glycol (PEG3) linker with a terminal aldehyde functional group used in PROTAC synthesis and general bioconjugation chemistry. It provides a short, flexible spacer (three ethylene glycol units) and an aldehyde reactive handle suitable for selective coupling reactions, enabling assembly of bifunctional molecules and small-molecule conjugates.
  • Provides an aldehyde functional handle for reductive amination and other aldehyde-specific couplings.
  • Offers a short, flexible PEG3 spacer for defined linker length.
  • High purity for research-grade applications.
  • Good solubility in common organic solvents.
  • Available in multiple pack sizes for flexible experimental needs.
Catalog No. 50-002-35035 Supplier Medchemexpress LLC Supplier No. HY124011100MG
m-PEG3-aldehyde is a monomethyl triethylene glycol (PEG3) linker with a terminal aldehyde functional group used in PROTAC synthesis and general bioconjugation chemistry. It provides a short, flexible spacer (three ethylene glycol units) and an aldehyde reactive handle suitable for selective coupling reactions, enabling assembly of bifunctional molecules and small-molecule conjugates.
  • Provides an aldehyde functional handle for reductive amination and other aldehyde-specific couplings.
  • Offers a short, flexible PEG3 spacer for defined linker length.
  • High purity for research-grade applications.
  • Good solubility in common organic solvents.
  • Available in multiple pack sizes for flexible experimental needs.