Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG3-aldehyde (stabilized with BHT) | 356066-46-1 | MFCD26910955 | 98.0% | 176.21 | C8H16O4 | 50MG
SDP

Catalog No. 5000235052
Encompass_Preferred

m-PEG3-aldehyde (stabilized with BHT) is a polyethylene glycol-based aldehyde linker used as a building block in the chemical synthesis of PROTACs and other bioconjugates. It provides a short PEG spacer with a reactive aldehyde terminus suitable for conjugation to hydrazide, aminooxy, or amine-functional groups.

  • Provides an aldehyde-terminated PEG3 spacer for conjugation.
  • Molecular formula C8H16O4 and molecular weight 176.21.
  • High purity (98.0%).
  • Appearance liquid, colorless to light yellow.
  • CAS number 356066-46-1.
  • Available in small milligram pack sizes for research use.
  • Recommended long-term storage at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-35052 Supplier Medchemexpress LLC Supplier No. HY124011A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG3-aldehyde (stabilized with BHT) is a polyethylene glycol-based aldehyde linker used as a building block in the chemical synthesis of PROTACs and other bioconjugates. It provides a short PEG spacer with a reactive aldehyde terminus suitable for conjugation to hydrazide, aminooxy, or amine-functional groups.

  • Provides an aldehyde-terminated PEG3 spacer for conjugation.
  • Molecular formula C8H16O4 and molecular weight 176.21.
  • High purity (98.0%).
  • Appearance liquid, colorless to light yellow.
  • CAS number 356066-46-1.
  • Available in small milligram pack sizes for research use.
  • Recommended long-term storage at -20°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.