Learn More
Medchemexpress LLC m-PEG3-aldehyde (stabilized with BHT) | 356066-46-1 | MFCD26910955 | 98.0% | 176.21 g/mol | C8H16O4 | 10MG
SDP
Catalog No. 5000235778
Encompass_Preferred
m-PEG3-aldehyde (stabilized with BHT) is a methoxy-terminated PEG3 aldehyde used as a PROTAC linker and synthetic building block for bioconjugation. The product is stabilized with butylated hydroxytoluene (BHT) and is supplied as a solid in milligram quantities for research use only. Molecular formula C8H16O4, molecular weight 176.21 g/mol.
  • Aldehyde functional group for facile conjugation.
  • Methoxy-terminated PEG3 spacer provides flexibility and solubility.
  • Stabilized with BHT to preserve aldehyde integrity during storage.
  • High reported purity of 98.0% suitable for synthetic applications.
  • Available in multiple small package sizes for laboratory-scale work.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-35778 Supplier Medchemexpress LLC Supplier No. HY124011A10MG
m-PEG3-aldehyde (stabilized with BHT) is a methoxy-terminated PEG3 aldehyde used as a PROTAC linker and synthetic building block for bioconjugation. The product is stabilized with butylated hydroxytoluene (BHT) and is supplied as a solid in milligram quantities for research use only. Molecular formula C8H16O4, molecular weight 176.21 g/mol.
  • Aldehyde functional group for facile conjugation.
  • Methoxy-terminated PEG3 spacer provides flexibility and solubility.
  • Stabilized with BHT to preserve aldehyde integrity during storage.
  • High reported purity of 98.0% suitable for synthetic applications.
  • Available in multiple small package sizes for laboratory-scale work.
  • Intended for research use only; not for clinical or human use.