Learn More
Medchemexpress LLC M-PEG3-aldehyde (stabilized with BHT) | 356066-46-1 | MFCD26910955 | 98.0% | 176.21 g/mol | C8H16O4 | 5MG
SDP
Catalog No. 5000238409
Encompass_Preferred
m-PEG3-aldehyde (stabilized with BHT) is a methoxy-terminated polyethylene glycol linker containing three ethylene glycol units and a terminal aldehyde. Provided for research use, it serves as a short, aldehyde-functionalized spacer for small-molecule conjugation and PROTAC synthesis; the BHT stabilizer helps reduce oxidation during storage and handling.
  • Terminal aldehyde enables oxime formation and reductive amination conjugation chemistries.
  • Short PEG3 spacer provides flexibility and improves solubility without large steric bulk.
  • Stabilized with BHT to limit oxidative degradation during storage.
  • High purity (98.0%) suitable for synthetic and medicinal chemistry applications.
  • Available in small milligram pack sizes for bench-scale workflows.
Catalog No. 50-002-38409 Supplier Medchemexpress LLC Supplier No. HY124011A5MG
m-PEG3-aldehyde (stabilized with BHT) is a methoxy-terminated polyethylene glycol linker containing three ethylene glycol units and a terminal aldehyde. Provided for research use, it serves as a short, aldehyde-functionalized spacer for small-molecule conjugation and PROTAC synthesis; the BHT stabilizer helps reduce oxidation during storage and handling.
  • Terminal aldehyde enables oxime formation and reductive amination conjugation chemistries.
  • Short PEG3 spacer provides flexibility and improves solubility without large steric bulk.
  • Stabilized with BHT to limit oxidative degradation during storage.
  • High purity (98.0%) suitable for synthetic and medicinal chemistry applications.
  • Available in small milligram pack sizes for bench-scale workflows.