Learn More
Medchemexpress LLC M-PEG3-Aminooxy | 248275-10-7 | 98.0% | C7H17NO4 | 1 G
SDP
Catalog No. 5000353220
Encompass_Preferred
m-PEG3-Aminooxy is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets the protein of interest. This product is intended strictly for research purposes and is not for patient use.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-53220 Supplier Medchemexpress LLC Supplier No. HY1403981G
m-PEG3-Aminooxy is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These compounds are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. PROTACs feature two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets the protein of interest. This product is intended strictly for research purposes and is not for patient use.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only