Learn More
Medchemexpress LLC m-PEG3-azide | 74654-06-1 | MFCD00020141 | >98% | C7H15N3O3 | 250 MG
SDP
Catalog No. 5000357258
Encompass_Preferred
m-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in various cycloaddition reactions.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups.
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
Catalog No. 50-003-57258 Supplier Medchemexpress LLC Supplier No. 250MGHY140827
m-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in various cycloaddition reactions.
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups.
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.