Learn More
Medchemexpress LLC m-PEG3-phosphonic ac 100mg
SDP
Catalog No. 5000205315
Encompass_Preferred
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05315 Supplier Medchemexpress LLC Supplier No. HYW096153100MG
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]