Learn More
Medchemexpress LLC m-PEG3-phosphonic ac 10mg
SDP
Catalog No. 5000204736
Encompass_Preferred
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04736 Supplier Medchemexpress LLC Supplier No. HYW09615310MG
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]