Learn More
Medchemexpress LLC m-PEG3-phosphonic ac 250mg
SDP
Catalog No. 5000204577
Encompass_Preferred
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04577 Supplier Medchemexpress LLC Supplier No. 250MGHYW096153
m-PEG3-phosphonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]