Learn More
Medchemexpress LLC m-PEG3-succinimidyl carbonate | 477775-77-2 | 99.4% | C12H19NO8 | 100 MG
SDP
Catalog No. 5000353132
Encompass_Preferred
m-PEG3-succinimidyl carbonate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • The other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53132 Supplier Medchemexpress LLC Supplier No. HY141112100MG
m-PEG3-succinimidyl carbonate is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • The other ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins