Learn More
Medchemexpress LLC M-PEG3-succinimidyl carbonate | 477775-77-2 | 99.4% | C12H19NO8 | 250 MG
SDP
Catalog No. 5000352563
Encompass_Preferred
m-PEG3-succinimidyl carbonate is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other to the target protein. These molecules function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-52563 Supplier Medchemexpress LLC Supplier No. 250MGHY141112
m-PEG3-succinimidyl carbonate is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other to the target protein. These molecules function by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only