Learn More
Medchemexpress LLC m-PEG4-Boc | 883554-11-8 | 292.37 g/mol | C14H28O6 | 250 MG
SDP
Catalog No. 5000202633
Encompass_Preferred
m-PEG4-Boc is a Boc-protected, cleavable polyethylene glycol linker containing four ethylene glycol units used for conjugation chemistry in ADC and PROTAC synthesis.
  • Cleavable Boc-protected PEG4 linker.
  • Designed for use in antibody-drug conjugate and PROTAC linker synthesis.
  • Provides a short, flexible spacer of four ethylene glycol units.
  • Available in multiple pack sizes, suitable for small-scale synthesis and development.
  • Supplied as an in-stock research reagent with associated product documentation.
Catalog No. 50-002-02633 Supplier Medchemexpress LLC Supplier No. HY141395250MG
m-PEG4-Boc is a Boc-protected, cleavable polyethylene glycol linker containing four ethylene glycol units used for conjugation chemistry in ADC and PROTAC synthesis.
  • Cleavable Boc-protected PEG4 linker.
  • Designed for use in antibody-drug conjugate and PROTAC linker synthesis.
  • Provides a short, flexible spacer of four ethylene glycol units.
  • Available in multiple pack sizes, suitable for small-scale synthesis and development.
  • Supplied as an in-stock research reagent with associated product documentation.