Learn More
Medchemexpress LLC m-PEG4-Br | 110429-45-3 | MFCD12545963 | 98.8% | 271.15 g/mol | C9H19BrO4 | 5 G
SDP
Catalog No. 5000203805
Encompass_Preferred
m-PEG4-Br is a methoxy-terminated PEG4 bromide linker (C9H19BrO4, MW 271.15 g/mol) used for bioconjugation chemistry, including antibody-drug conjugate and PROTAC linker synthesis. The linker provides a PEG4 spacer with a terminal bromide suitable for nucleophilic substitution and downstream functionalization.
  • Cleavable PEG4 linker with terminal bromide.
  • Suitable for ADC and PROTAC linker synthesis.
  • Provides increased hydrophilicity and spacer length.
  • High reported purity (approximately 98.8%).
  • Available in a 5 G pack size.
Catalog No. 50-002-03805 Supplier Medchemexpress LLC Supplier No. HY1301615G
m-PEG4-Br is a methoxy-terminated PEG4 bromide linker (C9H19BrO4, MW 271.15 g/mol) used for bioconjugation chemistry, including antibody-drug conjugate and PROTAC linker synthesis. The linker provides a PEG4 spacer with a terminal bromide suitable for nucleophilic substitution and downstream functionalization.
  • Cleavable PEG4 linker with terminal bromide.
  • Suitable for ADC and PROTAC linker synthesis.
  • Provides increased hydrophilicity and spacer length.
  • High reported purity (approximately 98.8%).
  • Available in a 5 G pack size.